Learn More
CCDC50, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CCDC50.
Brand: Abnova PAB20051.100uL
Description
This gene encodes a soluble, cytoplasmic, tyrosine-phosphorylated protein with multiple ubiquitin-interacting domains. Mutations in this gene cause nonsyndromic, postlingual, progressive sensorineural DFNA44 hearing loss. In mouse, the protein is expressed in the inner ear during development and postnatal maturation and associates with microtubule-based structures. This protein may also function as a negative regulator of NFkb signaling and as an effector of epidermal growth factor (EGF)-mediated cell signaling. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Sequence: QEKKDEDIARLLQEKELQEEKKRKKHFPEFPATRAYADSYYYEDGGMKPRVMKEAVSTPSRMAHRDQEWYDAEIARKLQEEELLATQVDMRAAQVAQDEEIARLLMAEEKKAYKKAKEREKSSLDKRKQDPEWKPKTAKAANSKSKESDESpecifications
| CCDC50 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CCDC50. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8IVM0 | |
| CCDC50 | |
| Recombinant protein corresponding to amino acids of human CCDC50. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:1000-1:2500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| CCDC50 | |
| C3orf6/DFNA44/YMER | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 152137 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only