Learn More
CC2D2A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CC2D2A.
Brand: Abnova PAB24295.100uL
Description
This gene encodes a coiled-coil and calcium binding domain protein that appears to play a critical role in cilia formation. Mutations in this gene cause Meckel syndrome type 6, as well as Joubert syndrome type 9. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: SVTPNDQCPRAEVSRREDVKKRSVYLKVLFNNKEVSRTVSRPLGADFRVHFGQIFNLQIVNWPESLTLQVYETVGHSSPTLLAEVFLPIPETTVVTGRAPTEEVEFSSNSpecifications
| CC2D2A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CC2D2A. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9P2K1 | |
| CC2D2A | |
| Recombinant protein corresponding to amino acids of human CC2D2A. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| CC2D2A | |
| JBTS9/KIAA1345/MKS6 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57545 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only