Learn More
CBL Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a synthetic peptide of CBL.
Brand: Abnova PAB0864.200ug
Description
The cbl oncogene was first identified as part of a transforming retrovirus which induces mouse pre-B and pro-B cell lymphomas. As an adaptor protein for receptor protein-tyrosine kinases, it positively regulates receptor protein-tyrosine kinase ubiquitination in a manner dependent upon its variant SH2 and RING finger domains. Ubiquitination of receptor protein-tyrosine kinases terminates signaling by marking active receptors for degradation. [provided by RefSeq
Specifications
| CBL | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a synthetic peptide of CBL. | |
| In PBS (0.2% gelatin, 0.09% sodium azide) | |
| C-CBL/CBL2/RNF55 | |
| Rabbit | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:200) The optimal working dilution should be determined by the end user. | |
| CBL | |
| CBL | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CBL. | |
| RUO | |
| 867 | |
| Store at 4°C. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only