Learn More
CASP7 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CASP7.
Brand: Abnova PAB13137.100ug
Description
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The precursor of this caspase is cleaved by caspase 3 and 10. It is activated upon cell death stimuli and induces apoptosis. Alternative splicing results in four transcript variants, encoding three distinct isoforms. [provided by RefSeq
Specifications
| CASP7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CASP7. | |
| In PBS (0.02% sodium azide) | |
| P55210 | |
| CASP7 | |
| A synthetic peptide corresponding to N-terminus 16 amino acids of human CASP7. | |
| RUO | |
| 840 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CASP7 | |
| CMH-1/ICE-LAP3/MCH3 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only