missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casp12 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Casp12.
Brand: Abnova PAB12908.100ug
This item is not returnable.
View return policy
Specifications
| Casp12 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Casp12. | |
| In PBS (0.02% sodium azide) | |
| CAA73532 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Caspase-12 (large) antibody can be used for the detection of both the propeptide and cleaved forms of caspase-12 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Casp12 | |
| Casp12 | |
| A synthetic peptide corresponding to mouse Casp12 large peptide. | |
| RUO | |
| 12364 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction