Learn More
CARS Rabbit anti-Human, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CARS.
Brand: Abnova PAB28563.100uL
Description
This gene encodes a class 1 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Specifications
| CARS | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CARS. | |
| In serum (0.05% sodium azide) | |
| CARS1/CYSRS/MGC:11246 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Rabbit | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot(1:10000) The optimal working dilution should be determined by the end user. | |
| CARS | |
| CARS | |
| Recombinant protein corresponding to N-term 6x His-human CARS. | |
| RUO | |
| 833 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only