Learn More
CARD9 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CARD9.
Brand: Abnova PAB5279.100ug
Description
The protein encoded by this gene is a member of the CARD protein family, which is defined by the presence of a characteristic caspase-associated recruitment domain (CARD). CARD is a protein interaction domain known to participate in activation or suppression of CARD containing members of the caspase family, and thus plays an important regulatory role in cell apoptosis. This protein was identified by its selective association with the CARD domain of BCL10, a postive regulator of apoptosis and NF-kappaB activation, and is thought to function as a molecular scaffold for the assembly of a BCL10 signaling complex that activates NF-kappaB. Several alternatively spliced transcript variants have been observed, but their full-length nature is not clearly defined. [provided by RefSeq
Specifications
| CARD9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CARD9. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| hCARD9 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CARD9 | |
| CARD9 | |
| A synthetic peptide corresponding to 3-24 amino acids at N-terminus of human CARD9. | |
| RUO | |
| 64170 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only