missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Canine IgG (H+L) Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB7316
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Canine | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:30000, ELISA 1:1000-1:30000(Primary) 1:100-1:500(for coating plates), Immunohistochemistry 1:200- 1:2000, Immunocytochemistry/Immunofluorescence 1:200- 1:2000, Immunohistochemistry-Paraffin 1:200- 1:2000 | |
| Dog IgG-heavy and light chain | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| By immunoelectrophoresis and ELISA this reacts specifically with dog IgG and with light chains common to other dog immunoglobulins. No was detected against non-immunoglobulin serum proteins. This may cross react with IgG from other species. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction