Learn More
CAMK2A/CAMK2B/CAMK2D (phospho T305), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CAMK2A/CAMK2B/CAMK2D.
Brand:  Abnova PAB29254.100uL
Product Code. 16224250
  • £349.00 / 100µL
Specifications
CAMK2A/CAMK2B/CAMK2D
Polyclonal
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CAMK2A/CAMK2B/CAMK2D.
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
CAMKA/KIAA0968
Rabbit
Affinity Chromatography
RUO
815/816/817
Human, Mouse, Rat
Liquid
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (1:50∼1:100) Western Blot (1:500∼1:1000) The optimal working dilution should be determined by the end user.
Q9UQM7
CAMK2A
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T305 of human CAMK2A/CAMK2B/CAMK2D.
100 μL
Primary
CAMK2A/CAMK2B/CAMK2D (phospho T305) polyclonal antibody detects endogenous levels of human CAMK2A/CAMK2B/CAMK2D only when phosphorylated at threonine 305.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.