Learn More
CAMK1D Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CAMK1D.
Brand: Abnova PAB2768.400uL
Description
This gene encodes a member of the Ca2+/calmodulin-dependent protein kinase 1 subfamily of serine/threonine kinases. The encoded protein may be involved in the regulation of granulocyte function through the chemokine signal transduction pathway. Alternatively spliced transcript variants encoding different isoforms of this gene have been described. [provided by RefSeq]
Specifications
| CAMK1D | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CAMK1D. | |
| In PBS (0.09% sodium azide) | |
| Q9HD31;NP_705718 | |
| CAMK1D | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CAMK1D. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| CAMK1D | |
| CKLiK/CaM-K1/CaMKID | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 57118 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only