missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cacng4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Cacng4.
Brand: Abnova PAB9886.100ug
This item is not returnable.
View return policy
Specifications
| Cacng4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Cacng4. | |
| In PBS (0.08% sodium azide) | |
| Cacng4 | |
| A synthetic peptide (conjugated with KLH) corresponding to rat Cacng4. | |
| RUO | |
| 140725 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Cacng4 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction