missing translation for 'onlineSavingsMsg'
Learn More

CACNA2D1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16183490
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16183490 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16183490 Supplier Abnova Supplier No. PAB20413.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against recombinant CACNA2D1.

This gene encodes a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq

Sequence: QPKNPKSQEPVTLDFLDAELENDIKVEIRNKMIDGESGEKTFRTLVKSQDERYIDKGNRTYTWTPVNGTDYSLALVLPTYSFYYIKAKLEETITQARSKKGKMKDSETLKPDNFEESGYTFIAPRDYCNDLKISDNNTEFL

Specifications

Antigen CACNA2D1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant CACNA2D1.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene CACNA2D1
Gene Accession No. P54289
Gene Alias CACNA2/CACNL2A/CCHL2A/MHS3
Gene Symbols CACNA2D1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human CACNA2D1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 781
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.