Learn More
CABP5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CABP5.
Brand: Abnova PAB24053.100uL
Description
The product of this gene belongs to a subfamily of calcium binding proteins, which share similarity to calmodulin. Calcium binding proteins are an important component of calcium mediated cellular signal transduction. Expression of this gene is retina-specific. The mouse homolog of this protein has been shown to express in the inner nuclear layer of the retina, suggested its role in neuronal functioning. The specific function of this gene is unknown. Study of the transcripts and genomic structure revealed that the 5' end of this gene is complementary and reverse to that of CABP3 gene, and the sequence beyond the overlapping region is nearly identical to that of CABP3. Thus, these two genes encode the protein products with distinct N-terminal halves but identical C-terminal halves. [provided by RefSeq]
Sequence: ITLVELQQAMQRLLGERLTPREISEVVREADVNGDGSpecifications
| CABP5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CABP5. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9NP86 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 56344 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| CABP5 | |
| CABP5 | |
| Recombinant protein corresponding to amino acids of human CABP5. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only