missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant C7.
Brand: Abnova PAB28437.100uL
This item is not returnable.
View return policy
Description
C7 is a component of the complement system. It participates in the formation of Membrane Attack Complex (MAC). People with C7 deficiency are prone to bacterial infection. [provided by RefSeq
Sequence: SAFLCGSSLKWSPEMKNARCVQKENPLTQAVPKCQRWEKLQNSRCVCKMPYECGPSLDVCAQDERSKRILPLTVCKMHVLHCQGRNYTLTGRDSCTLPASAEKACGACPLWGKCDAESSKCVCREASECEEEGFSICVEVNGKEQTMSECSpecifications
| C7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant C7. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| P10643 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 730 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry(1:50-1:200) The optimal working dilution should be determined by the end user. | |
| C7 | |
| C7 | |
| Recombinant protein corresponding to amino acids of recombinant C7. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction