Learn More
C6orf108 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant C6orf108.
Brand: Abnova PAB22327.100uL
Description
This gene was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation. Two alternative transcripts encoding different proteins have been described. [provided by RefSeq
Sequence: MAAAMVPGRSESWERGEPGRPALYFCGSIRGGREDRTLYERIVSRLRRFGTVLTEHVSpecifications
| C6orf108 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant C6orf108. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| O43598 | |
| C6orf108 | |
| Recombinant protein corresponding to amino acids of human C6orf108. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| C6orf108 | |
| RCL/RP3-330M21.3/dJ330M21.3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10591 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only