missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5AR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of C5AR1.
Brand: Abnova PAB18321.100ug
This item is not returnable.
View return policy
Specifications
| C5AR1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| C5AR1 | |
| C5A/C5AR/C5R1/CD88 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 728 | |
| Human | |
| Liquid |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of C5AR1. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| P21730 | |
| C5AR1 | |
| A synthetic peptide corresponding to residues surrounding S338 of human C5AR1. | |
| 100 μg | |
| Primary | |
| This antibody is specific to C5AR1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction