missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native C4.
Brand: Abnova PAB5003.150ug
This item is not returnable.
View return policy
Specifications
| C4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native C4. | |
| In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
| C4A | |
| Native purified human C4. | |
| RUO | |
| 720/721 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| C4/C4A2/C4A3/C4A4/C4A6/C4S/CO4/CPAMD2/MGC164979/RG | |
| Rabbit | |
| 150 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction