missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf40 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of C2orf40.
Brand: Abnova PAB25734.100ug
This item is not returnable.
View return policy
Specifications
| C2orf40 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunocytochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| C2orf40 | |
| ECRG4 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 84417 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of C2orf40. | |
| In PBS (0.02% sodium azide) | |
| AAI09386 | |
| C2orf40 | |
| A synthetic peptide corresponding to 20 amino acids at C-terminus of human C2orf40. | |
| 100 μg | |
| Primary | |
| ECRG4 antibody is predicted to not cross-react with other ECRG family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction