missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1R, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant C1R.
Brand: Abnova PAB28388.100uL
This item is not returnable.
View return policy
Description
Sequence: PNNFETTTVITVPTGYRVKLVFQQFDLEPSEGCFYDYVKISADKKSLGRFCGQLGSPLGNPPGKKEFMSQGNKMLLTFHTDFSNEENGTIMFYKGFLAYYQAVDLDECASRSKSGEEDPQPQCQHLCSpecifications
| C1R | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant C1R. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| D3DUT5 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 715 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:250) Immunofluorescence (1-4 ug/ml) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| C1R | |
| C1R | |
| Recombinant protein corresponding to amino acids of human C1R. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido