Learn More
C17orf39, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant C17orf39.
Brand: Abnova PAB24319.100uL
Description
The multiprotein Mediator complex is a coactivator required for activation of RNA polymerase II transcription by DNA bound transcription factors. The protein encoded by this gene is thought to be a subunit of the Mediator complex. This gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq
Sequence: DTGNSYLCGYLKIKGLTEEYPTLTTFFEGEIISKKHPFLTRKWDADEDVDRKHWGKFLAFYQYAKSFNSDDFDYESpecifications
| C17orf39 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant C17orf39. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8IVV7 | |
| C17orf39 | |
| Recombinant protein corresponding to amino acids of human C17orf39. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| C17orf39 | |
| MGC3048 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 79018 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only