Learn More
BTG2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BTG2.
Brand: Abnova PAB28556.100uL
Description
The protein encoded by this gene is a member of the BTG/Tob family. This family has structurally related proteins that appear to have antiproliferative properties. This encoded protein is involved in the regulation of the G1/S transition of the cell cycle. [provided by RefSeq
Sequence: QEALTEHYKHHWFPEKPSKGSGYRCIRINHKMDPIISRVASQIGLSQPQLHQLLPSELTLWVDPYEVSYRIGEDGSICVLYEEAPLAASCGLLTCKNQVLLGSpecifications
| BTG2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BTG2. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| P78543 | |
| BTG2 | |
| Recombinant protein corresponding to amino acids of recombinant BTG2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:200-1:500) Western Blot(1:250-1:500) The optimal working dilution should be determined by the end user. | |
| BTG2 | |
| MGC126063/MGC126064/PC3/TIS21 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7832 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only