Learn More
BTG1 Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BTG1.
Brand: Abnova PAB5273.100ug
Description
This gene is a member of an anti-proliferative gene family that regulates cell growth and differentiation. Expression of this gene is highest in the G0/G1 phases of the cell cycle and downregulated when cells progressed through G1. The encoded protein interacts with several nuclear receptors, and functions as a coactivator of cell differentiation. This locus has been shown to be involved in a t(8;12)(q24;q22) chromosomal translocation in a case of B-cell chronic lymphocytic leukemia. [provided by RefSeq
Specifications
| BTG1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BTG1. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| BTG1 | |
| A synthetic peptide corresponding to N-terminus of human BTG1. | |
| RUO | |
| 694 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| BTG1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Chicken, Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only