Learn More
BRS3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BRS3.
Brand: Abnova PAB24983.100uL
Description
Mammalian bombesin-like peptides (see MIM 137260) are widely distributed in the central nervous system as well as in the gastrointestinal tract, where they modulate smooth-muscle contraction, exocrine and endocrine processes, metabolism, and behavior. They bind to G protein-coupled receptors on the cell surface to elicit their effects. Bombesin-like peptide receptors include gastrin-releasing peptide receptor (MIM 305670), neuromedin B receptor (MIM 162341), and bombesin-like receptor-3 (BRS3) (Ohki-Hamazaki et al., 1997).[supplied by OMIM
Specifications
| BRS3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BRS3. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| BRS3 | |
| A synthetic peptide corresponding to BRS3. | |
| 100 μL | |
| Primary | |
| BRS3 polyclonal antibody detects endogenous levels of BRS3 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| BRS3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 680 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only