Learn More
BRS3 Rabbit anti-Bovine, Dog, Guinea pig, Hamster, Horse, Human, Monkey, Mouse, Rabbit, Rat, Sheep, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BRS3.
Marke: Abnova PAB16381.50ug
Beschreibung
Mammalian bombesin-like peptides (see MIM 137260) are widely distributed in the central nervous system as well as in the gastrointestinal tract, where they modulate smooth-muscle contraction, exocrine and endocrine processes, metabolism, and behavior. They bind to G protein-coupled receptors on the cell surface to elicit their effects. Bombesin-like peptide receptors include gastrin-releasing peptide receptor (MIM 305670), neuromedin B receptor (MIM 162341), and bombesin-like receptor-3 (BRS3) (Ohki-Hamazaki et al., 1997).[supplied by OMIM
Spezifikation
| BRS3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BRS3. | |
| In PBS (0.09% sodium azide) | |
| P32247 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 680 | |
| Bovine, Canine, Equine, Guinea Pig, Hamster, Human, Monkey, Mouse, Rabbit, Rat, Sheep | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| BRS3 | |
| BRS3 | |
| A synthetic peptide (conjugated with KLH) corresponding to human BRS3. | |
| 50 μg | |
| Primary | |
| 1st Cytoplasmic domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only