Learn More
BRIP1 (phospho S990), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BRIP1.
Brand: Abnova PAB15895.100ug
Description
The protein encoded by this gene is a member of the RecQ DEAH helicase family and interacts with the BRCT repeats of breast cancer, type 1 (BRCA1). The bound complex is important in the normal double-strand break repair function of breast cancer, type 1 (BRCA1). This gene may be a target of germline cancer-inducing mutations. [provided by RefSeq
Specifications
| BRIP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of BRIP1. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| BACH1/FANCJ/FLJ90232/MGC126521/MGC126523/OF | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| BRIP1 | |
| BRIP1 | |
| Synthetic phosphopeptide corresponding to residues surrounding S990 of human BRIP1. | |
| RUO | |
| 83990 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only