Learn More
BRD1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BRD1.
Brand: Abnova PAB20028.100uL
Beskrivelse
This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types. [provided by RefSeq
Sequence: IAAEVGQSSMWISTDAAASVLEPLKVVWAKCSGYPSYPALIIDPKMPRVPGHHNGVTIPAPPLDVLKIGEHMQTKSDEKLFLVLFFDNKRSWQWLPKSKMVPLGIDETIDKLKMMEGRNSSIRKAVRIAFDRAMNHLSRVHGEPTSDTekniske data
| BRD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BRD1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| O95696 | |
| BRD1 | |
| Recombinant protein corresponding to amino acids of human BRD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| BRD1 | |
| BRL/BRPF1/BRPF2/DKFZp686F0325 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23774 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
For Research Use Only