Learn More
BRAP, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BRAP.
Brand: Abnova PAB27873.100uL
Description
The protein encoded by this gene was identified by its ability to bind to the nuclear localization signal of BRCA1 and other proteins. It is a cytoplasmic protein which may regulate nuclear targeting by retaining proteins with a nuclear localization signal in the cytoplasm. [provided by RefSeq
Sequence: SLVVIRLELAEHSPVPAGFGFSAAAGEMSDEEIKKTTLASAVACLEGKSPGEKVAIIHQHLGRREMTDVIIETMKSNPDELKTTVEERKSSEASpecifications
| BRAP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BRAP. | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| BRAP2/IMP/RNF52 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8315 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:100-1:250) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| BRAP | |
| BRAP | |
| Recombinant protein corresponding to amino acids of human BRAP. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only