missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Bovine IgA/Secretory Component Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69292
This item is not returnable.
View return policy
Description
IgA/Secretory Component Polyclonal antibody specifically detects IgA/Secretory Component in Bovine samples. It is validated for ELISA, Immunodiffusion, Immunoelectrophoresis
Specifications
| IgA/Secretory Component | |
| Polyclonal | |
| Unconjugated | |
| Delipidated, heat inactivated lyophilized whole antiserum | |
| Highly purified IgA/Secretory Component isolated from pooled bovine milk. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Secondary | |
| Bovine | |
| Lyophilized |
| ELISA, Immunodiffusion, Immunoelectrophoresis | |
| LYOPH | |
| ELISA, Immunodiffusion, Immunoelectrophoresis | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction