Learn More
BOK, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BOK.
Brand: Abnova PAB25702.100ug
Description
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains all four BCL-2 like domains (BH1, 2, 3 and 4) and is a pro-apoptotic BCL-2 protein identified in the ovary. [provided by RefSeq
Specifications
| BOK | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| BOK | |
| BCL2L9/BOKL/MGC4631 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 666 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of BOK. | |
| In PBS (0.05% sodium azide) | |
| AAH06203 | |
| BOK | |
| A synthetic peptide corresponding to 16 amino acids at N-terminus of human BOK. | |
| 100 μg | |
| Primary | |
| At least three isoforms of BOK are known to exist; this antibody will not detect the smallest isoform. BOK antibody is predicted to not cross-react with other Bcl-2 protein family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only