Learn More
BMP15 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BMP15.
Brand: Abnova PAB27235.100ug
Description
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The transforming growth factor-beta superfamily includes large families of growth and differentiation factors. It is thought that this protein may be involved in oocyte maturation and follicular development as a homodimer or by forming heterodimers with a related protein, Gdf9. Defects in this gene are the cause of ovarian dysgenesis 2
Specifications
| BMP15 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| BMP15 | |
| GDF9B/ODG2/POF4 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 9210 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of BMP15. | |
| In PBS (0.02% sodium azide) | |
| NP_005439 | |
| BMP15 | |
| A synthetic peptide corresponding to 18 amino acids at N-terminus of human BMP15. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only