Learn More
BLOC1S1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BLOC1S1.
Brand: Abnova PAB21399.100uL
Description
BLOC1S1 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules (Starcevic and Dell'Angelica, 2004 [PubMed 15102850]).[supplied by OMIM
Sequence: LLKEHQAKQNERKELQEKRRREAITAATCLTEALVDHLNVGVAQAYMNQRKLDHEVKTLQVQAAQFAKQTGQWIGMVENFNQALKEIGDVENWARSIELDMRTIATALEYVYKGQSpecifications
| BLOC1S1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BLOC1S1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P78537 | |
| BLOC1S1 | |
| Recombinant protein corresponding to amino acids of human BLOC1S1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| BLOC1S1 | |
| BLOS1/FLJ39337/FLJ97089/GCN5L1/MGC87455/MICoA/RT14 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2647 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only