Learn More
BICD2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BICD2.
Brand: Abnova PAB13388.100ug
Description
This gene is one of two human homologs of Drosophila bicaudal-D and a member of the Bicoid family. It has been implicated in dynein-mediated, minus end-directed motility along microtubules. It has also been reported to be a phosphorylation target of NIMA related kinase 8. Two alternative splice variants have been described. [provided by RefSeq
Specifications
| BICD2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BICD2. | |
| In PBS (0.02% sodium azide) | |
| CAI41013 | |
| BICD2 | |
| A synthetic peptide corresponding to internal region 14 amino acids of human BICD2. | |
| RUO | |
| 23299 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| BICD2 | |
| KIAA0699/bA526D8.1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only