Learn More
BICD2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BICD2.
Brand: Abnova PAB21698.100uL
Description
This gene is one of two human homologs of Drosophila bicaudal-D and a member of the Bicoid family. It has been implicated in dynein-mediated, minus end-directed motility along microtubules. It has also been reported to be a phosphorylation target of NIMA related kinase 8. Two alternative splice variants have been described. [provided by RefSeq
Sequence: EQKNSLRKELSHYMSINDSFYTSHLHVSLDGLKFSDDAAEPNNDAEALVNGFEHGGLAKLPLDNKTSTPKKEGLAPPSPSLVSDLLSELNISESpecifications
| BICD2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BICD2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8TD16 | |
| BICD2 | |
| Recombinant protein corresponding to amino acids of human BICD2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| BICD2 | |
| KIAA0699/bA526D8.1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23299 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only