Learn More
BHLHE40, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant BHLHE40.
Brand: Abnova PAB12512.100uL
Description
This gene encodes a basic helix-loop-helix protein expressed in various tissues. Expression in the chondrocytes is responsive to the addition of Bt2cAMP. The encoded protein is believed to be involved in the control of cell differentiation. [provided by RefSeq
Specifications
| BHLHE40 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant BHLHE40. | |
| In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
| NP_003661.1 | |
| BHLHE40 | |
| Recombinant protein corresponding to C-terminus of human BHLHE40. | |
| RUO | |
| 8553 | |
| Store at 4°C. Do not freeze. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| BHLHE40 | |
| BHLHB2/DEC1/FLJ99214/SHARP-2/STRA13/Stra14 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only