Learn More
BFSP2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BFSP2.
Brand: Abnova PAB23290.100uL
Description
More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, the protein product of this gene (phakinin), and filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene have been associated with juvenile-onset, progressive cataracts and Dowling-Meara epidermolysis bullosa simplex. [provided by RefSeq
Sequence: LSRNYEEDVKLLHKQLAGCELEQMDAPIGTGLDDILETIRIQWERDVEKNRVEAGALLQAKQQAEVAHMSQTQEEKLAAALRVESpecifications
| BFSP2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BFSP2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q13515 | |
| BFSP2 | |
| Recombinant protein corresponding to amino acids of human BFSP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| BFSP2 | |
| CP47/CP49/LIFL-L/MGC142078/MGC142080/PHAKOSIN | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8419 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only