Learn More
BDKRB2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BDKRB2.
Brand: Abnova PAB26164.50ug
Description
This gene encodes a receptor for bradykinin. The 9 aa bradykinin peptide elicits many responses including vasodilation, edema, smooth muscle spasm and pain fiber stimulation. This receptor associates with G proteins that stimulate a phosphatidylinositol-calcium second messenger system. Alternate start codons result in two isoforms of the protein. [provided by RefSeq
Specifications
| BDKRB2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BDKRB2. | |
| In PBS (0.09% sodium azide) | |
| P30411 | |
| BDKRB2 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human BDKRB2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except BAHD1 (44%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15-30 ug/mL) The optimal working dilution should be determined by the end user. | |
| BDKRB2 | |
| B2R/BK-2/BK2/BKR2/BRB2/DKFZp686O088 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 624 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only