missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCO2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BCO2.
Brand: Abnova PAB23586.100uL
This item is not returnable.
View return policy
Description
BCDO2 catalyzes the asymmetric oxidative cleavage of beta-carotene in carotene metabolism (Kiefer et al., 2001 [PubMed 11278918]).[supplied by OMIM
Sequence: VDFLPVMVHRLPVFKRYMGNTPQKKAVFGQCRGLPCVAPLLTTVEEAPRGISARVWGHFPKWLNGSLLRIGPGKFEFGKDKYNHWSpecifications
| BCO2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BCO2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9BYV7 | |
| BCO2 | |
| Recombinant protein corresponding to amino acids of human BCO2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| BCO2 | |
| B-DIOX-II/BCDO2/FLJ34464 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 83875 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction