Learn More
BCLAF1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BCLAF1.
Brand: Abnova PAB21902.100uL
Description
This gene encodes a transcriptional repressor that interacts with several members of the BCL2 family of proteins. Overexpression of this protein induces apoptosis, which can be suppressed by co-expression of BCL2 proteins. The protein localizes to dot-like structures throughout the nucleus, and redistributes to a zone near the nuclear envelope in cells undergoing apoptosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: QKSPEIHRRIDISPSTLRKHTRLAGEERVFKEENQKGDKKLRCDSADLRHDIDRRRKERSKERGDSKGSRESSGSRKQEKTPKDYKEYKSYKDDSKHKREQDHSRSSSSSASPSSPSSRSpecifications
| BCLAF1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BCLAF1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9NYF8 | |
| BCLAF1 | |
| Recombinant protein corresponding to amino acids of human BCLAF1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| BCLAF1 | |
| BTF/KIAA0164/bK211L9.1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9774 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only