Learn More
BCHE, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant BCHE.
Brand: Abnova PAB28449.100uL
Description
Mutant alleles at the BCHE locus are responsible for suxamethonium sensitivity. Homozygous persons sustain prolonged apnea after administration of the muscle relaxant suxamethonium in connection with surgical anesthesia. The activity of pseudocholinesterase in the serum is low and its substrate behavior is atypical. In the absence of the relaxant, the homozygote is at no known disadvantage. [provided by RefSeq
Sequence: QQLALQWVQKNIAAFGGNPKSVTLFGESAGAASVSLHLLSPGSHSLFTRAILQSGSFNAPWAVTSLYEARNRTLNLAKLTGCSRENETEIIKCLRNKDPQEILLNEAFVVPYGTPLSVNFGPTVDSpecifications
| BCHE | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant BCHE. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| F8WEX7 | |
| BCHE | |
| Recombinant protein corresponding to amino acids of recombinant BCHE. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry(1:50-1:200) The optimal working dilution should be determined by the end user. | |
| BCHE | |
| CHE1/E1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 590 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only