missing translation for 'onlineSavingsMsg'
Learn More

Bcar1 (phospho Y762), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of Bcar1.

Brand:  Abnova PAB7934.100uL

Product Code. 16080748

  • £483.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Bcar1
Polyclonal
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Bcar1.
In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Cas/Crkas/P130CAS
Rabbit
100 μL
Primary
This rat sequence is the same as Tyr-664 in human p130 Cas and Tyr-668 in mouse p130 Cas.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunoprecipitation, Western Blot
Unconjugated
ELISA (1:2000) Immunoprecipitation (5-10 ul) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Bcar1
Bcar1
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y762 of rat Bcar1.
RUO
25414
Human, Mouse, Rat
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only