missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bax, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant Bax.
Brand: Abnova PAB14668.50uL
This item is not returnable.
View return policy
Specifications
| Bax | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant Bax. | |
| In serum (0.05% sodium azide) | |
| NP_620120 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Canine, Gerbil, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:5000) Immunoprecipitation (1:50-1:200) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| Bax | |
| Bax | |
| Recombinant protein corresponding to full length mouse Bax. | |
| RUO | |
| 12028 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction