Learn More
BAK1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BAK1.
Brand: Abnova PAB27022.100uL
Description
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form oligomers or heterodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein localizes to mitochondria, and functions to induce apoptosis. It interacts with and accelerates the opening of the mitochondrial voltage-dependent anion channel, which leads to a loss in membrane potential and the release of cytochrome c. This protein also interacts with the tumor suppressor P53 after exposure to cell stress. [provided by RefSeq
Specifications
| BAK1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| BAK1 | |
| BAK/BAK-LIKE/BCL2L7/CDN1/MGC117255/MGC3887 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 578 | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of BAK1. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| Q16611 | |
| BAK1 | |
| A synthetic peptide corresponding to human BAK1. | |
| 100 μL | |
| Primary | |
| BAK1 polyclonal antibody detects endogenous levels of BAK1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only