Learn More
BAIAP3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BAIAP3.
Brand: Abnova PAB13345.100ug
Description
This p53-target gene encodes a brain-specific angiogenesis inhibitor. The protein is a seven-span transmembrane protein and a member of the secretin receptor family. It interacts with the cytoplasmic region of brain-specific angiogenesis inhibitor 1. This protein also contains two C2 domains, which are often found in proteins involved in signal transduction or membrane trafficking. Its expression pattern and similarity to other proteins suggest that it may be involved in synaptic functions. [provided by RefSeq
Specifications
| BAIAP3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BAIAP3. | |
| In PBS (0.02% sodium azide) | |
| EAW85671 | |
| BAIAP3 | |
| A synthetic peptide corresponding to N-terminus 13 amino acids from of human BAIAP3. | |
| RUO | |
| 8938 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| BAIAP3 | |
| BAP3/KIAA0734/MGC138334 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only