Learn More
AURKB Rabbit anti-Bovine, Chimpanzee, Dog, Human, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AURKB.
Brand: Abnova PAB10025.100ug
Description
Chromosomal segregation during mitosis as well as meiosis is regulated by kinases and phosphatases. The Aurora kinases associate with microtubules during chromosome movement and segregation. Aurora kinase B localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers, and Aurora kinase A (MIM 603072) localizes to centrosomes (Lampson et al., 2004 [PubMed 14767480]).[supplied by OMIM
Specifications
| AURKB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AURKB. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Q96GD4;NP_004208 | |
| AURKB | |
| A synthetic peptide corresponding to amino acids 227-238 of human AURKB. | |
| RUO | |
| 9212 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| ELISA (1:5000-1:20000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| AURKB | |
| AIK2/AIM-1/AIM1/ARK2/AurB/IPL1/STK12/STK5 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Bovine, Canine, Chimpanzee, Human, Mouse, Pig, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only