Learn More
AURKA Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant AURKA.
Brand: Abnova PAB28676.100uL
Description
The protein encoded by this gene is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression. A processed pseudogene of this gene has been found on chromosome 1, and an unprocessed pseudogene has been found on chromosome 10. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: ISGPVKATAPVGGPKRVLVTQQFPCQNPLPVNSGQAQRVLCPSNSSQRVPLQAQKLVSSHKPVQNQKQKQLQATSVPHPVSRPLNNTQKSKQPLPSAPENNPEEELASKQKNEESKKRQWALEDFEIGRPLGKGKFGNVYLSpecifications
| AURKA | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant AURKA. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| AIK/ARK1/AURA/AURORA2/BTAK/MGC34538/STK15/STK6/STK7 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6790 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| AURKA | |
| AURKA | |
| Recombinant protein corresponding to amino acids of human AURKA. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only