Learn More
ATP7A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ATP7A.
Brand: Abnova PAB20632.100uL
Description
This gene encodes a transmembrane protein that functions in copper transport across membranes. The protein localizes to the trans-Golgi network, where it is predicted to supply copper to copper-dependent enzymes in the secretory pathway. The protein relocalizes to the plasma membrane under conditions of elevated extracellular copper and functions in the efflux of copper from cells. Mutations in this gene result in Menkes disease, X-linked cutis laxa, and occipital horn syndrome. [provided by RefSeq
Sequence: TETLGTCIDFQVVPGCGISCKVTNIEGLLHKNNWNIEDNNIKNASLVQIDASNEQSSTSSSMIIDAQISNALNAQQYKVLIGNREWMIRNGLVINNDVNDFMTEHERKGRTAVLVAVDDELCSpecifications
| ATP7A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ATP7A. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q04656 | |
| ATP7A | |
| Recombinant protein corresponding to amino acids of human ATP7A. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| ATP7A | |
| FLJ17790/MK/MNK | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 538 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only