Learn More
ATP6V0A4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ATP6V0A4.
Brand: Abnova PAB21000.100uL
Description
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c″, and d. This gene is one of four genes in man and mouse that encode different isoforms of the a subunit. Alternatively spliced transcript variants encoding the same protein have been described. Mutations in this gene are associated with renal tubular acidosis associated with preserved hearing. [provided by RefSeq]
Sequence: RASHRKSQLQASRIQEDATENIEGDSSSPSSRSGQRTSADTHGALDDHGEEFNFGDVFVHQAIHTIEYCLGCISNTASSpecifications
| ATP6V0A4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ATP6V0A4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9HBG4 | |
| ATP6V0A4 | |
| Recombinant protein corresponding to amino acids of human ATP6V0A4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| ATP6V0A4 | |
| A4/ATP6N1B/ATP6N2/MGC130016/MGC130017/RDRTA2/RTA1C/RTADR/STV1/VPH1/VPP2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 50617 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only