Learn More
ATP5B Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ATP5B.
Brand: Abnova PAB28448.100uL
Description
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the beta subunit of the catalytic core. [provided by RefSeq
Sequence: TSPSPKAGAATGRIVAVIGAVVDVQFDEGLPPILNALEVQGRETRLVLEVAQHLGESTVRTIAMDGTEGLVRGQKVLDSGAPIKIPVGPETLGRIMNVIGEPIDERGPIKTKQFAPIHAEAPEFMEMSVEQEILVTGIKVVDSpecifications
| ATP5B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ATP5B. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| F8VPV9 | |
| ATP5B | |
| Recombinant protein corresponding to amino acids of recombinant ATP5B. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:500-1:1000) Western Blot(1:250-1:500) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| ATP5B | |
| ATPMB/ATPSB/MGC5231 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 506 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only