Learn More
ATP2C1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ATP2C1.
Brand: Abnova PAB19368.100ug
Description
The protein encoded by this gene belongs to the family of P-type cation transport ATPases. This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of the calcium. Defects in this gene cause Hailey-Hailey disease, an autosomal dominant disorder. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Specifications
| ATP2C1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ATP2C1 | |
| ATP2C1A/BCPM/HHD/KIAA1347/PMR1/SPCA1/hSPCA1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 27032 | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ATP2C1. | |
| In PBS (0.02% sodium azide) | |
| NP_001001486 | |
| ATP2C1 | |
| A synthetic peptide corresponding to 19 amino acids near C-terminus of human ATP2C1. | |
| 100 μg | |
| Primary | |
| At least four isoforms of ATP2C1 are known to exist; this antibody will recognize only the three longest isoforms. ATP2C1 antibody will not cross-react with ATP2C2. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only