missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ATG8.
Brand: Abnova PAB11359.500ug
This item is not returnable.
View return policy
Specifications
| ATG8 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ATG8. | |
| In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| NP_009475 | |
| ATG8 | |
| Recombinant protein corresponding to Saccharomyces cerevisiae ATG8. | |
| RUO | |
| 852200 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:20000-1:100000) Western Blot (1:4000-1:8000) The optimal working dilution should be determined by the end user. | |
| ATG8 | |
| APG8/AUT7/CVT5 | |
| Rabbit | |
| 500 μg | |
| Primary | |
| Yeast | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction